missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL, p Thr357 Antibody (954724), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB91871DL594
This item is not returnable.
View return policy
Description
MLKL Monoclonal antibody specifically detects MLKL in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| MLKL | |
| Monoclonal | |
| DyLight 594 | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Synthetic peptide from human MLKL phosphorylated at T357 [UniProt Q8NB16] | |
| 0.1 mL | |
| Necroptosis, Protein Kinase | |
| 197259 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 954724 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction