missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL, p Thr357 Antibody (954724), DyLight 755, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB91871IR
This item is not returnable.
View return policy
Description
MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow Cytometry (Negative).
Specifications
| MLKL | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MLKL | |
| Synthetic peptide from human MLKL phosphorylated at T357 [UniProt Q8NB16] | |
| 100 μL | |
| Protein Kinase | |
| 197259 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry | |
| 954724 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow Cytometry (Negative), Immunocytochemistry | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction