missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MLL3 Antibody (7D5D7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 287.00 - € 511.00
Specifications
| Antigen | MLL3 |
|---|---|
| Clone | 7D5D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025786
|
Novus Biologicals™
NBP3-27146-0.025ml |
0.025 mL |
€ 287.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30035602
|
Novus Biologicals™
NBP3-27146-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLL3 Monoclonal antibody specifically detects MLL3 in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| MLL3 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| ALR-like protein, DKFZp686C08112, EC 2.1.1.43, FLJ12625, HALRMGC119851, histone-lysine N-methyltransferase MLL3, histone-lysine N-methyltransferase, H3 lysine-4 specific, Homologous to ALR protein, KIAA1506FLJ38309, KMT2C, KMT2CMGC119852, Lysine N-methyltransferase 2C, MGC119853, myeloid/lymphoid or mixed-lineage leukemia 3, Myeloid/lymphoid or mixed-lineage leukemia protein 3 | |
| Purified recombinant fragment of human MLL3 (AA: 1-205) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 7D5D7 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 58508 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title