missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLPH Mouse anti-Human, Clone: OTI6E3, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808433S
This item is not returnable.
View return policy
Description
MLPH Monoclonal antibody specifically detects MLPH in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MLPH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BV36 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79083 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E3 | |
| Unconjugated | |
| Mlph | |
| 2210418F23Rik; 5031433I09Rik; AW228792; D1Wsu84e; Exophilin-3; l(1)-3Rk; l1Rk3; leaden; leaden protein; Ln; Melanophilin; MGC2771; MGC59733; Mlph; OTTMUSP00000025451; SLAC2A; Slac-2a; SlaC2-a; Slp homolog lacking C2 domains a; synaptotagmin-like protein 2a | |
| Human recombinant protein fragment corresponding to amino acids 295-600 of human MLPH produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction