missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLPH Mouse anti-Human, Clone: OTI6E3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808433
This item is not returnable.
View return policy
Description
Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.Specifications
| MLPH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BV36 | |
| MLPH | |
| Human recombit protein fragment corresponding to amino acids 295-600 of human MLPH produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E3 | |
| Unconjugated | |
| MLPH | |
| SLAC2-A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79083 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction