missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLPH Polyclonal Antibody
GREENER_CHOICE

Product Code. 17177893
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177893 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177893 Supplier Invitrogen™ Supplier No. PA5109965

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EQLPLQYLAD VDTSDEESIR AHVMASHHSK RRGRASSESQ IFELNKHISA VECLLTYLEN TVVPPLAKGL GAGVRTEADV EEEALRRKLE ELTSNVSDQE TSSEEEEAKD EKAEPNRDKS VGPLPQADPE VGTAAHQTNR QEKSPQDPGD PVQYNRTTDE ELSELEDRVA VTASEVQQAE SEVSDIESRI AALRAAGLTV KPSGKPRRKS NLPIFLPRVA GKLGKRPEDP NADPSSEAKA MAVPYLLRRK FSNSLKSQGK DDDSFDRKSV YRGSLTQRNP NARKGMASHT FAKPVVAHQS.

Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLPH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mlph
Gene Accession No. Q91V27, Q9BV36
Gene Alias 2210418F23Rik; 5031433I09Rik; AW228792; D1Wsu84e; Exophilin-3; l(1)-3Rk; l1Rk3; leaden; leaden protein; Ln; Melanophilin; MGC2771; MGC59733; Mlph; OTTMUSP00000025451; SLAC2A; Slac-2a; SlaC2-a; Slp homolog lacking C2 domains a; synaptotagmin-like protein 2a
Gene Symbols Mlph
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 301-600 of human MLPH (NP_077006.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171531, 316620, 79083
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.