missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MLSTD2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553585

Product Code. 15761521

  • € 580.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: YYEGKNVLLT GATGFLGKVL LEKLLRSCPK VNSVYVLVRQ KAGQTPQERV EEVLSGKLFD RLRDENPDFR EKIIAINSEL TQPKLALSEE DKEVIIDSTN IIFHCAATVR FNENLRDAVQ LNVIATRQLI LLAQQMKNLE VFM Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

MLSTD2 catalyzes the reduction of saturated fatty acyl-CoA with chain length C16 or C18 to fatty alcohols.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLSTD2
Polyclonal
Unconjugated
FAR1
2600011M19Rik; 2900034E22Rik; 3732409C05Rik; AI850429; FAR1; fatty acyl CoA reductase 1; fatty acyl-CoA reductase 1; male sterility domain containing 2; male sterility domain-containing protein 2; MLSTD2; PFCRD; SDR10E1; short chain dehydrogenase/reductase family 10E, member 1; UNQ2423/PRO4981
Rabbit
Antigen affinity chromatography
RUO
84188
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WVX9
FAR1
Recombinant protein corresponding to Human MLSTD2. Recombinant protein control fragment (Product #RP-90598).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.