missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLXIP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81965
This item is not returnable.
View return policy
Description
MLXIP Polyclonal specifically detects MLXIP in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MLXIP | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| MLXIP | |
| MLXIP antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human MLXIP. Amino Acid Squence: SPHREHPPKGYDFDTVNK | |
| Affinity Purified | |
| RUO | |
| 22877 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 ug/mL, ELISA | |
| bHLHe36BHLHE36, Class E basic helix-loop-helix protein 36, KIAA0867MondoA, MIRMLX-interacting protein, MLX interacting protein, MONDOAMlx interactor, Transcriptional activator MondoA | |
| Rabbit | |
| 21 kDa | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction