missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLXIP Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81965H
This item is not returnable.
View return policy
Description
MLXIP Polyclonal antibody specifically detects MLXIP in Human samples. It is validated for Western Blot, ELISA
Specifications
| MLXIP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 22877 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| HRP | |
| bHLHe36BHLHE36, Class E basic helix-loop-helix protein 36, KIAA0867MondoA, MIRMLX-interacting protein, MLX interacting protein, MONDOAMlx interactor, Transcriptional activator MondoA | |
| MLXIP antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human MLXIP. Amino Acid Squence: SPHREHPPKGYDFDTVNK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction