Learn More
Invitrogen™ MMAA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140091
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ERGHKLSVLA VDPSSCTSGG SLLGDKTRMT ELSRDMNAYI RPSPTRGTLG GVTRTTNEAI LLCEGAGYDI ILIETVGVGQ SEFAVAD Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 93%.
The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
Specifications
| MMAA | |
| Polyclonal | |
| Unconjugated | |
| MMAA | |
| 2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVH4 | |
| MMAA | |
| Recombinant protein corresponding to Human MMAA. Recombinant protein control fragment (Product #RP-108955). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.