missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMAB Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502167
This item is not returnable.
View return policy
Description
This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group.Specifications
| MMAB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EY8 | |
| MMAB | |
| Full length human recombit protein of human MMAB produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3B8 | |
| Unconjugated | |
| MMAB | |
| ATR, CFAP23, cblB, cob | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 326625 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction