missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMACHC Antibody (OTI4E9), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72734MFV450
This item is not returnable.
View return policy
Description
MMACHC Monoclonal antibody specifically detects MMACHC in Human samples. It is validated for Flow Cytometry
Specifications
| MMACHC | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI4E9 | |
| Flow Cytometry | |
| cblC, DKFZP564I122, FLJ25671, methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria, methylmalonic aciduria and homocystinuria type C protein, RP11-291L19.3 | |
| Full length human recombinant protein of human MMACHC(NP_056321) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 25974 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction