missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMACHC Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504935
This item is not returnable.
View return policy
Description
The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Specifications
| MMACHC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y4U1 | |
| MMACHC | |
| Full length human recombit protein of human MMACHC produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1B7 | |
| Unconjugated | |
| MMACHC | |
| cblC | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25974 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction