missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMADHC Antibody (OTI1G4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72739FR
This item is not returnable.
View return policy
Description
MMADHC Monoclonal specifically detects MMADHC in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MMADHC | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| MMADHC | |
| Recombinant protein fragment corresponding to amino acids 39-296 of human MMADHC (NP_056517) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1G4 | |
| Western Blot | |
| cblD, chromosome 2 open reading frame 25, methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria, mitochondrial, protein C2orf25, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 27249.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction