Learn More
MMAF-d8 hydrochloride, MedChemExpress
Product Code. 16471529
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471529 1 mg 1mg
16449207 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16471529 Supplier Medchem Express Supplier No. HY15579AS
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
MedChemExpress D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Specifications
Chemical Name or Material MMAF-d8 hydrochloride
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C39H58D8ClN5O8
Technique Cancer-programmed Cell Death
Color Off-White
Solubility Information DMSO : 100 mg/mL (128.79 mM; Need ultrasonic)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl
Molecular Weight (g/mol) 776.47
Quantity 1 mg
Formula Weight 776.47
Percent Purity 98.97%
Grade Research
Show More Show Less
Research purposes only