Learn More
MME Monoclonal Antibody (OTI2A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810614S
Encompass_Preferred
 View more versions of this product
Product Code. 17593904
  • € 242.00 / 30µL
Description
MME Monoclonal antibody specifically detects MME in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
MME
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P98177
Mouse
Affinity chromatography
RUO
4303
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2A4
Unconjugated
FOXO4
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase
Human recombinant protein fragment corresponding to amino acids 206-505 of human FOXO4 produced in E.coli.
30 μL