Learn More
MME Mouse anti-Human, Clone: , Abnova™
Mouse monoclonal antibody raised against human MME.
Brand: Abnova MAB12519.50uL
Description
This gene encodes a common acute lymphocytic leukemia antigen that is an important cell surface marker in the diagnosis of human acute lymphocytic leukemia (ALL). This protein is present on leukemic cells of pre-B phenotype, which represent 85% of cases of ALL. This protein is not restricted to leukemic cells, however, and is found on a variety of normal tissues. It is a glycoprotein that is particularly abundant in kidney, where it is present on the brush border of proximal tubules and on glomerular epithelium. The protein is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon, enkephalins, substance P, neurotensin, oxytocin, and bradykinin. This gene, which encodes a 100-kD type II transmembrane glycoprotein, exists in a single copy of greater than 45 kb. The 5' untranslated region of this gene is alternatively spliced, resulting in four separate mRNA transcripts. The coding region is not affected by alternative splicing. [provided by RefSeq]
Specifications
| MME | |
| Monoclonal | |
| Mouse monoclonal antibody raised against human MME. | |
| In PBS, pH 7.4 (0.03% sodium azide). | |
| MME | |
| Recombinant protein corresponding to human MME. | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| ELISA (1:10000) Immunohistochemistry (Frozen sections) (1:200-1:400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| CALLA/CD10/DKFZp686O16152/MGC126681/MGC126707/NEP | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4311 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.