missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MME Mouse anti-Human, Clone: HI10a, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13777.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human MME.
- High specificity
- High reactivity
Specifications
| MME | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| P08473 | |
| MME | |
| Acute CALLA leukemia blast cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| HI10a | |
| Mouse monoclonal antibody raised against human MME. | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| CALLA/CD10/DKFZp686O16152/MGC126681/MGC126707/NEP | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4311 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction