missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-14/MT1-MMP Antibody (128527), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9181PECY55
This item is not returnable.
View return policy
Description
MMP-14/MT1-MMP Monoclonal antibody specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Flow Cytometry
Specifications
| MMP-14/MT1-MMP | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 128527 | |
| Flow Cytometry | |
| EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-14/MT1-MMP, Tyr112-Ala541 (predicted), Accession # P50281 | |
| 0.1 mL | |
| Cancer, Extracellular Matrix | |
| 4323 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction