missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-2 Antibody (2C1) - (Pro and Active) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80853
This item is not returnable.
View return policy
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| MMP-2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2-5 ug/ml, Flow Cytometry, ELISA : 1 ug/ml, Immunohistochemistry : 5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30681163), Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin 5ug/ml. Use reported in scientific literature (PMID 24828425), Immunohistochemistry-Frozen : 5 ug/ml, CyTOF-ready | |
| 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Guinea Pig | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| 2C1 | |
| Unconjugated | |
| PBS | |
| MMP2 | |
| Activated recombinant human MMP2. [UniProt# P08253] | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors | |
| 4313 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction