missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-24/MT5-MMP Antibody (143908), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB924C
This item is not returnable.
View return policy
Description
MMP-24/MT5-MMP Monoclonal antibody specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| MMP-24/MT5-MMP | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10893 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 143908 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
| E. coli-derived recombinant mouse MMP-24/MT5-MMP, Arg125-Gln296, Accession # Q9R0S2 | |
| 0.5 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction