missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-24/MT5-MMP Antibody (RM0289-7J71), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11803PE
This item is not returnable.
View return policy
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Mouse samples. It is validated for Flow Cytometry.
Specifications
| MMP-24/MT5-MMP | |
| Monoclonal | |
| PE | |
| EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 10893 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| RM0289-7J71 | |
| Flow Cytometry | |
| MMP24 | |
| Mouse recombinant MMP-24/MT5-MMP (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| This antibody weakly cross reacts with mouse MMP3 and MMP-9. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction