missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-3 Antibody (MMP3/1994R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08627IR
This item is not returnable.
View return policy
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| MMP-3 | |
| Monoclonal | |
| DyLight 755 | |
| CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
| Recombinant fragment (around aa 317-327) of human MMP-3 protein (Uniprot: P08254) | |
| 100 μg | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 4314 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| MMP3/1994R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction