missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-8 Antibody (100608), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9081L
This item is not returnable.
View return policy
Description
MMP-8 Monoclonal antibody specifically detects MMP-8 in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
Specifications
| MMP-8 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 100608 | |
| ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-8, Phe21-Gly467, Accession # AAZ38714 | |
| 0.5 mL | |
| Cancer | |
| 4317 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction