missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-8 Antibody (206), DyLight 550 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89361R
This item is not returnable.
View return policy
Description
MMP-8 Monoclonal antibody specifically detects MMP-8 in Human samples. It is validated for ELISA, ELISA
Specifications
| MMP-8 | |
| Monoclonal | |
| DyLight 550 | |
| CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-8 (NP_002415.1; Met1-Gly467). | |
| 0.1 mL | |
| Cancer | |
| 4317 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 206 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction