Learn More
MMP-8 Antibody (MM0023-7A11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB110-60989PE
Product Code. 18561173
  • € 607.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
MMP-8 Monoclonal antibody specifically detects MMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
RUO
Primary
Human
PBS
Cancer
4317
Store at 4C in the dark.