missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (ITGAX/1243), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54432C
This item is not returnable.
View return policy
Description
CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| CD11c | |
| Monoclonal | |
| DyLight 650 | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Recombinant fragment (aa 637-827) of human CD11c protein (exact sequence is proprietary) (Uniprot: P20702) | |
| 100 μg | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| ITGAX/1243 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction