missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RPL35A Antibody, Novus Biologicals™

Product Code. 30046366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30046366 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30046366 Supplier Novus Biologicals™ Supplier No. NBP330234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RPL35A Polyclonal antibody specifically detects RPL35A in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL35A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias Cell growth-inhibiting gene 33 protein, DBA5,60S ribosomal protein L35a, ribosomal protein L35a
Host Species Rabbit
Immunogen Between 60 and 110
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6165
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.