missing translation for 'onlineSavingsMsg'
Learn More

MMP-9 Antibody (S51-82), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

€ 184.00 - € 614.00

Specifications

Antigen MMP-9
Clone S51-82
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18068524
View Documents
Novus Biologicals
NBP2-59699
100 μg
€ 614.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18607268
View Documents
Novus Biologicals
NBP2-59699-25ug
25 μg
€ 184.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

MMP-9
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
MMP9
IgG2a
Protein G purified
S51-82
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Human, Mouse, Rat
4318
Fusion protein amino acids 1-708 (full length) of rat MMP9
Primary
Detecs ∼92kDa and ∼82kDa (pro and active forms).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.