missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17878372
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17878372 200 μL 200µL
17773274 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17878372 Supplier Invitrogen™ Supplier No. PA5119271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guinea Pig Polyclonal Antibody

MMP13 Polyclonal Antibody for Western Blot, ICC/IF

MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP13
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MMP13
Gene Accession No. O62806
Gene Alias cb1034; Clg; CLG3; Collagenase; collagenase 3; collagenase 3a; matrix metalloproteinase 13a; collagenase-1; Collagenase3; Collagenase-3; collagenase-3 precursor; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase-13; matrix metalloproteinase-13; MMP-13; interstitial protein; MMP; Mmp1; MMP13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13
Gene Symbols MMP13
Host Species Guinea Pig
Immunogen Recombinant protein Matrix Metalloproteinase 13. The antigen corresponds to amino acid range 70-245 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008685
Target Species Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.