missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP14 Recombinant Mouse Monoclonal Antibody (LEM-2/15)
GREENER_CHOICE

Product Code. 17981891
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981891 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981891 Supplier Invitrogen™ Supplier No. MA547931

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Specificity: This antibody binds to residues 218-233 of the V-B loop of MT1-MMP with a Kd of 2.32E10-9M.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP14
Applications ELISA, Inhibition Assays, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone LEM-2/15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene MMP14
Gene Accession No. P50281
Gene Alias AI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); matrix metalloproteinase-14; matrix metalloproteinase-14a; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alpha
Gene Symbols MMP14
Host Species Mouse
Immunogen The cyclic peptide possessing the sequence of residues 218-233 of the V-B-loop of MT1-MMP.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.