missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP16 Polyclonal Antibody

Description
Recommended positive controls: HepG2, mouse kidney, rat kidney, Rat dorsal root ganglion. Predicted reactivity: Mouse (98%), Rat (98%), Japanese Medaka (83%), Xenopus laevis (92%), Chicken (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Matrix metallopeptidase 16 during embryonic development, reproduction, and tissue remodeling. MMP-16 is also involved in the ECM degradation processes associated with arthritis and metastasis. In humans, MMP-16 is a part of a cluster of MMP genes at the 11q22.3 locus.
Specifications
Specifications
| Antigen | MMP16 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Mmp16 |
| Gene Accession No. | O35548, P51512, Q9WTR0 |
| Gene Alias | C8orf57; DKFZp761D112; matrix metallo protease; matrix metallopeptidase 16; matrix metallopeptidase 16 (membrane-inserted); matrix metalloproteinase 16; matrix metalloproteinase-16; Membrane type 3-MMP; membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase; MMP; MMP-1; Mmp16; MMP-16; MMPs; MMPX2; MMP-X2; Mt3mmp; MT3-MMP; MT-MMP 3; MT-MMP2; MTMMP3; MT-MMP3; Putative transmembrane protein C8orf57 |
| Gene Symbols | Mmp16 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?