Learn More
Invitrogen™ MMP19 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA513187
Product Code. 10400300
  • € 578.00 / 400µL
Description
MMP19 Polyclonal Antibody for Western Blot, IHC (P), Flow
MMP-19 (RASI-1, MMP-18) is a 508 amino acid peptide, originally isolated as an autoantigen from the synovium of a patient suffering from rheumatoid arthritis (RA). Its presence on the surface of activated PBMCs, TH1 lymphocytes, and Jurkat T lymphoma cells in the synovium of acute RA patients, suggests that MMP-19 plays a role in RA-associated joint tissue destruction. MMP-19 exists in the smooth muscle cells of the tunica media of large blood vessels of the RA patient, but not in the endothelial cell layer. Acutely inflamed tissue synovial capillaries strongly express MMP-19 in the cytoplasm. Regulated induction of MMP-19 in capillary endothelial cells during acute inflammation suggest a role in angiogenesis. MMP-19 is a single 2.7kb transcript found in mammary gland, placenta, lung, pancreas, ovary, small intestine, spleen, thymus, prostate, testis, colon, and heart.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMP19
Polyclonal
Unconjugated
MMP19
CODA; matrix metallo protease; matrix metallopeptidase 19; matrix metalloproteinase 19; matrix metalloproteinase RASI; Matrix metalloproteinase-18; matrix metalloproteinase-19; matrix metalloproteinase-beta19; MMP; MMP18; MMP-18; Mmp19; MMP-19; MMPs; Rasi; RASI-1
Rabbit
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
RUO
4327
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99542
MMP19
KLH conjugated synthetic peptide between 344-373 amino acids from the C-terminal region of human MMP19.
400 μL