Learn More
MMP24 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
Antigen MMP24
Concentration 450 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233208
View Documents
Proteintech
30102-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
30237843
View Documents
Proteintech
30102-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Description
Description
The MMP24 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MMP24. This antibody recognizes human, mouse, rat antigen. The MMP24 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MMP24
Western Blot, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
450 μg/mL
Polyclonal
Liquid
RUO
10893
IgG
Antigen affinity purification
Polyclonal Antibody