missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Monoclonal Antibody (OTI3C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806915
This item is not returnable.
View return policy
Description
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
| MMP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08254 | |
| MMP3 | |
| Human recombit protein fragment corresponding to amino acids 246-477 of human MMP3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3C6 | |
| Unconjugated | |
| MMP3 | |
| CHDS6, MMP-3, SL-1, STMY, STMY1, STR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4314 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction