missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Monoclonal Antibody (OTI3C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806915S
This item is not returnable.
View return policy
Description
MMP3 Monoclonal antibody specifically detects MMP3 in Human samples. It is validated for Western BlotSpecifications
| MMP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35916 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2324 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C6 | |
| Unconjugated | |
| FLT4 | |
| CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
| Human recombinant protein fragment corresponding to amino acids 1063-1363 of human FLT4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?