missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP9 Recombinant Rabbit Monoclonal Antibody (043), Biotin, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 163.00 - € 510.00
Specifications
| Antigen | MMP9 |
|---|---|
| Clone | 043 |
| Concentration | 0.5 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019988
|
Invitrogen™
MA552185 |
20 μL |
€ 163.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019879
![]() |
Invitrogen™
MA552184 |
100 μg |
€ 510.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP9 Recombinant Monoclonal antibody specifically detects MMP9 in Human samples. It is validated for ELISASpecifications
| MMP9 | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 67 kDa matrix metalloproteinase-9; 82 kDa matrix metalloproteinase-9; 92 kDa gelatinase; 92 kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; 92-kDa type IV collagenase; AW743869; B/MMP9; Clg4b; collagenase type IVB; fj05a08; Gel B; gelatinase B; Gelatinase-B; GELB; macrophage gelatinase; MANDP2; matrix metallo protease; matrix metallopeptidase 9; matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92kDa matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase9; matrix metalloproteinase-9; MMP; Mmp9; MMP-9; mmp9 protein; MMP-9 protein; MMPs; Preproform of 92-kDa type IV collagenase (MMP-9): N-terminal; pro-MMP-9; type IV collagenase; type V collagenase; wu:fb02g06; wu:fb07b05; wu:fi98c09; wu:fj05a08; ZFMMP-9; zgc:64165 | |
| Recombinant Human MMP-9 protein (Ala 20-Asp707). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MMP9 |
| 043 | |
| ELISA | |
| Biotin | |
| Rabbit | |
| Human | |
| P14780 | |
| 4318 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title