missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMR/CD206/Mannose Receptor Antibody (685641), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25342APCCY7
This item is not returnable.
View return policy
Description
MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Flow Cytometry
Specifications
| MMR/CD206/Mannose Receptor | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 685641 | |
| Flow Cytometry | |
| CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206, Leu19-Lys1383 (Thr399Ala) and (Leu407Phe), Accession # P22897 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 4360 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction