missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MNAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576700
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
| MNAR | |
| Polyclonal | |
| Unconjugated | |
| PELP1 | |
| 4930563C04Rik; HMX3; Mnar; modulator of nongenomic activity of estrogen receptor; Modulator of non-genomic activity of estrogen receptor; P160; PELP1; proline and glutamic acid rich nuclear protein; proline, glutamate and leucine rich protein 1; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; RGD1306320; transcription factor HMX3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27043, 360552, 75273 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q56B11, Q8IZL8, Q9DBD5 | |
| PELP1 | |
| Recombinant full length Human MNAR. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction