missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 540.75
Specifications
| Antigen | MNAT1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MNAT1 Polyclonal specifically detects MNAT1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MNAT1 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4331 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDELESSDLPVALLLAQHKDRSTQLEMQLEKPKPVKPVTFSTGIKMGQHISLAPIHKLEEALYEYQPLQIETYGPHVPELEMLGRLGYLNHVRAASPQDLAG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CAP35, CDK7/cyclin-H assembly factor, cyclin G1 interacting protein, Cyclin-G1-interacting protein, MAT1TFB3, Menage a trois, menage a trois 1 (CAK assembly factor), menage a trois homolog 1, cyclin H assembly factor (Xenopus laevis), p35, p36, RING finger protein 66, RING finger protein MAT1, RNF66CDK-activating kinase assembly factor MAT1 | |
| MNAT1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title