missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNDA Antibody (OTI4H6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72741CL1
This item is not returnable.
View return policy
Description
MNDA Monoclonal antibody specifically detects MNDA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MNDA | |
| Monoclonal | |
| CoraFluor 1 | |
| myeloid cell nuclear differentiation antigen, PYHIN3 | |
| Human recombinant protein fragment corresponding to amino acids 37-321 of human MNDA(NP_002423) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 4332 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| OTI4H6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction