Learn More
Cbl-c Antibody (01), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06621F
Product Code. 18665248
  • € 423.00 / 0.10mL
Description
Cbl-c Monoclonal antibody specifically detects Cbl-c in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cbl-c
Monoclonal
FITC
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA
01
ELISA
Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cbl-c (AAH28915.1; Met1-Ala474).
0.1 mL
Stem Cell Markers
23624
Store at 4°C in the dark.