missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNX1/HLXB9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99434-100ul
This item is not returnable.
View return policy
Description
MNX1/HLXB9 Polyclonal antibody specifically detects MNX1/HLXB9 in Mouse samples. It is validated for Western Blot
Specifications
| MNX1/HLXB9 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MNX1/HLXB9 | |
| 100 μg | |
| Cell Biology, Neuroscience, Stem Cells | |
| 3110 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction