missing translation for 'onlineSavingsMsg'
Learn More

MNX1/HLXB9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 369.00 - € 570.00

Specifications

Antigen MNX1/HLXB9
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18206322
View Documents
Novus Biologicals
NBP2-57703
100 μL
€ 570.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601108
View Documents
Novus Biologicals
NBP2-57703-25ul
25 μL
€ 369.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MNX1/HLXB9 Polyclonal specifically detects MNX1/HLXB9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MNX1/HLXB9
Polyclonal
Rabbit
Human
3110
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NFRIDALLAVDPPRAASAQSAPLALVTSLAAAASGTGG
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9
MNX1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.