missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598902
This item is not returnable.
View return policy
Description
MOB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
The protein encoded by this gene is a component of the Hippo signaling pathway, which controls organ size and tumor growth by enhancing apoptosis. Loss of the encoded protein results in cell proliferation and cancer formation. The encoded protein is also involved in the control of microtubule stability during cytokinesis. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| MOB1 | |
| Polyclonal | |
| Unconjugated | |
| Mob1b | |
| 1110003E08Rik; 4022402H07Rik; AU015450; B230364F10; C2orf6; MATS1; MATS2; MOB kinase activator 1A; MOB kinase activator 1B; MOB1; Mob1 alpha; mob1 homolog 1A; Mob1 homolog 1B; MOB1 Mps One Binder homolog A; MOB1 Mps One Binder homolog B; MOB1, Mps One Binder kinase activator-like 1A; MOB1, Mps One Binder kinase activator-like 1B; mob1A; Mob1b; MOB4A; MOB4B; Mobk1b; MOBKL1A; MOBKL1B; MOBKL1B/MOB4A; mps one binder kinase activator-like 1A; Mps one binder kinase activator-like 1B; Protein Mob4B | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55233, 92597 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7L9L4, Q9H8S9 | |
| MOB1A, Mob1b | |
| Recombinant Human MOB kinase activator 1A protein (2-216AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction