Learn More
Invitrogen™ MOB4 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5142709
Product Code. 17856432
  • € 465.00 / 100µL
Description
Additional Information: This antibody is expected to recognize all three reported isoforms (NP_056202.2; NP_955776.1; NP_001094289.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
This gene was identified based on its similarity with the mouse counterpart. Studies of the mouse counterpart suggest that the expression of this gene may be regulated during oocyte maturation and preimplantation following zygotic gene activation. Alternatively spliced transcript variants encoding distinct isoforms have been observed. Naturally occurring read-through transcription occurs between this locus and the neighboring locus HSPE1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MOB4
Polyclonal
Unconjugated
MOB4
2610109B12Rik; 2C4D; CGI-95; Class II mMOB1; MOB family member 4, phocein; MOB1; Mob1 homolog 3; MOB1, Mps One Binder kinase activator-like 3; mob3; MOB4; Mobkl1; Mobkl3; MOB-like protein phocein; mps one binder kinase activator-like 3; Phocein; phocein, Mob-like protein; PHOCN; PREI3; Preimplantation protein 3
Goat
Ammonium Sulfate Precipitation
RUO
171050, 19070, 25843
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q6PEB6, Q9QYW3, Q9Y3A3
MOB4
Peptide with sequence C-NSNKYFPSRVSIKES (aa 137-151).