Learn More
MOB4A Antibody (OTI3G9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72744C
Product Code. 18154096
  • € 751.00 / 0.10mL
Description
MOB4A Monoclonal specifically detects MOB4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MOB4A
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
MOB1B
Full length recombinant protein of human MOB4A (NP_775739) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry
OTI3G9
Western Blot, Immunohistochemistry
MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A
Mouse
Affinity Purified
Signal Transduction
92597.0
Store at 4°C in the dark.