Learn More
MOB4A/MOBKL1B Antibody (01), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06512MFV610
This item is not returnable.
View return policy
Description
MOB4A/MOBKL1B Monoclonal antibody specifically detects MOB4A/MOBKL1B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| MOB4A/MOBKL1B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 92597 | |
| Store at 4C in the dark. | |
| IgG2a |
| Immunocytochemistry | |
| 01 | |
| Immunocytochemistry/ Immunofluorescence | |
| ATS2;MATS2;MOB kinase activator 1B;MOB1B;MOB4A;MOBKL1A | |
| This antibody was obtained from a mouse immunized with purified, recombinant Human MOB4A/MOBKL1B (Uniprot#: Q7L9L4; Met1-Arg216). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |