Learn More
Manutan Mobile lift mini-table, strength: 150 kg, overallwidth: 450 mm
Brand:  Manutan A036646
Encompass_Preferred
Mobile lift mini-table, strength: 150 kg, overallwidth: 450 mm
Product Code. 15692782
  • € 1051.49 / Each
Only null left
Estimated Shipment: 09-03-2026