Learn More
Knick Elektronische™ Mobile pH Meter
Product Code. 15301097
Click to view available options
Dimensions (L x W x H):
132 x 156 x 30 mm
Unit Size:
Each
Product Code. 15301097
Brand: Knick Elektronische™ 902PH
Only null left
Add to basket
Add to basket
The Portavo 902 pH Meter comes with:
  • 4 AA batteries and pre-mounted quiver
  • Carrying strap
  • Quickstart instructions in various languages
  • Specific test report
  • Safety instructions
  • Data carrier with detailed user manuals
Specifications
Includes pH Meter, 4 AA batteries, Pre-mounted quiver, Carrying strap, Quickstart instructions, Specific test report, Data carrier with detailed user manuals
Range (pH) -2 to 16
Range (mV) 1300 to 1300
Temperature Range (Metric) 20°C to 120°C
Memory Yes
Inputs DIN 19262
IP Rating IP66/67 with pressure compensation
Dimensions (L x W x H) 132 x 156 x 30 mm
Weight (Metric) 500 g
Warranty 3 Years
Keypad Yes
Material PA12 GF30 + TPE
Model Portavo 902 pH
Relative Humidity Range 0 to 95%, short-term condensing allowed
Type Mobile pH Meter